ENAH5802EABA Enamine

methyl 1-(dimethyl-S-aminosulfonimidoyl)piperidine-4-carboxylate hydrochloride

  • MDL number MFCD31924319
  • Cas Number : 2344678-18-6
  • Molecular Weight : 285.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7355398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355398
Availability details for
Quantity
  • Availability
  • Add to Cart