ENAH5802EABC Enamine

methyl 1-phenyl-4,5,6,7-tetrahydro-1H-indazole-6-carboxylate

  • MDL number MFCD32198453
  • Cas Number : 2113470-18-9
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 1-phenyl-4,5,6,7-tetrahydro-1H-indazole-6-carboxylate
Inchi Code 1S/C15H16N2O2/c1-19-15(18)11-7-8-12-10-16-17(14(12)9-11)13-5-3-2-4-6-13/h2-6,10-11H,7-9H2,1H3
Inchi Key HGEQBNMSCMNYLS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7443449

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7443449
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart