ENAH5802EABF Enamine

6-ethyl 3-methyl 5H,6H,7H,8H-pyrido[4,3-c]pyridazine-3,6-dicarboxylate

  • Cas Number : 2375268-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7440701

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440701
Availability details for
Quantity
  • Availability
  • Add to Cart