ENAH5802EAC3 Enamine

methyl 2-{[(benzyloxy)carbonyl]amino}-2-(oxetan-3-yl)acetate

  • MDL number MFCD14586431
  • Cas Number : 1255717-04-4
  • Molecular Weight : 279.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl {[(benzyloxy)carbonyl]amino}(3-oxetanyl)acetate
Inchi Code 1S/C14H17NO5/c1-18-13(16)12(11-8-19-9-11)15-14(17)20-7-10-5-3-2-4-6-10/h2-6,11-12H,7-9H2,1H3,(H,15,17)
Inchi Key CPWNUOJLLPXDNI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7437109

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7437109
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart