ENAH5802EAC9 Enamine

tert-butyl 2-(3-bromophenyl)-2-cyanoacetate

  • MDL number MFCD32066349
  • Cas Number : 1107634-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431026

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431026
Availability details for
Quantity
  • Availability
  • Add to Cart