ENAH5802EACB Enamine

methyl 2-{4-[(tert-butyldimethylsilyl)oxy]-3-methoxyphenyl}acetate

  • MDL number MFCD32065900
  • Cas Number : 2361644-00-8
  • Molecular Weight : 310.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-(4-((tert-butyldimethylsilyl)oxy)-3-methoxyphenyl)acetate
Inchi Code 1S/C16H26O4Si/c1-16(2,3)21(6,7)20-13-9-8-12(10-14(13)18-4)11-15(17)19-5/h8-10H,11H2,1-7H3
Inchi Key IIVHTCHOHGGTKN-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430323
Availability details for
Quantity
  • Availability
  • Add to Cart