ENAH5802EAD3 Enamine

2-tert-butyl 3-methyl 8-imino-8-oxo-8lambda6-thia-2-azaspiro[4.5]decane-2,3-dicarboxylate

  • MDL number MFCD32066564
  • Cas Number : 2361645-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739574
Availability details for
Quantity
  • Availability
  • Add to Cart