ENAH5802EAD6 Enamine

N-hydroxy-2-phenylethanecarbonimidoyl chloride

  • MDL number MFCD28011381
  • Cas Number : 1086251-93-5
  • Molecular Weight : 169.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-chloro-2-nitrosoethyl)benzene
Inchi Code 1S/C8H8ClNO/c9-8(10-11)6-7-4-2-1-3-5-7/h1-5,8H,6H2
Inchi Key PGYNQHNWIXFADP-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6494438

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6494438
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart