ENAH5802EADD Enamine

5-chloro-1-methyl-1H-indole-2-carbohydrazide

  • Cas Number : 162081-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443397
Availability details for
Quantity
  • Availability
  • Add to Cart