ENAH5802EADE Enamine

N'-(2-methylquinolin-4-yl)-3-phenylpropanehydrazide hydrochloride

  • MDL number MFCD32066644
  • Cas Number : 2361635-80-3
  • Molecular Weight : 341.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N'-(2-methylquinolin-4-yl)-3-phenylpropanehydrazide hydrochloride
Inchi Code 1S/C19H19N3O.ClH/c1-14-13-18(16-9-5-6-10-17(16)20-14)21-22-19(23)12-11-15-7-3-2-4-8-15;/h2-10,13H,11-12H2,1H3,(H,20,21)(H,22,23);1H
Inchi Key GRRYJUXRTQCGJR-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7432987

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7432987
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart