ENAH5802EAE0 Enamine

2-(2-{[(furan-2-yl)methyl]imino}-3-(3-methoxyphenyl)-2,3-dihydro-1,3-thiazol-4-yl)acetohydrazide

  • Cas Number : 556006-97-4
  • Molecular Weight : 358.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06389
Availability details for
Quantity
  • Availability
  • Add to Cart