ENAH5802EAE7 Enamine

4-hydrazinyl-6-methoxypyrimidine

  • MDL number MFCD09863255
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-hydrazino-6-methoxypyrimidine
Inchi Code 1S/C5H8N4O/c1-10-5-2-4(9-6)7-3-8-5/h2-3H,6H2,1H3,(H,7,8,9)
Inchi Key IEVYUPUJSDLOID-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-275886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-275886
Availability details for
Quantity
  • Availability
  • Add to Cart