ENAH5802EAE8 Enamine

4,4-dimethyl-1,4-azasilinan-1-amine dihydrochloride

  • MDL number MFCD32198159
  • Cas Number : 2375273-53-1
  • Molecular Weight : 217.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,4-dimethyl-1,4-azasilinan-1-amine dihydrochloride

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7444662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444662
Availability details for
Quantity
  • Availability
  • Add to Cart