ENAH5802EAEE Enamine

2-hydrazinyl-3-nitro-5-(trifluoromethyl)pyridine

  • MDL number MFCD01566807
  • Cas Number : 288246-61-7
  • Molecular Weight : 222.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-hydrazino-3-nitro-5-(trifluoromethyl)pyridine
Inchi Code 1S/C6H5F3N4O2/c7-6(8,9)3-1-4(13(14)15)5(12-10)11-2-3/h1-2H,10H2,(H,11,12)
Inchi Key CUUQSZLMISKOHS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-4309216

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-4309216
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart