ENAH5802EAF7 Enamine

3-(4-methylphenyl)-1,2-oxazolidine

  • Cas Number : 1780260-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7439389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7439389
Availability details for
Quantity
  • Availability
  • Add to Cart