ENAH5802EAF8 Enamine

O-decylhydroxylamine

  • MDL number MFCD21920915
  • Cas Number : 29812-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1851870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1851870
Availability details for
Quantity
  • Availability
  • Add to Cart