ENAH5802EAFB Enamine

amino 4-methylbenzene-1-sulfonate

  • MDL number MFCD16876702
  • Cas Number : 52913-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-41
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-137607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137607
Availability details for
Quantity
  • Availability
  • Add to Cart