ENAH5802EAFE Enamine

4-isocyanato-1,2-thiazole

  • MDL number MFCD32066319
  • Cas Number : 1538994-91-0
  • Molecular Weight : 126.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-isocyanatoisothiazole
Inchi Code 1S/C4H2N2OS/c7-3-5-4-1-6-8-2-4/h1-2H
Inchi Key XNMRCWNUIBXZDF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-2488746

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P285,P301+P312,P302+P352,P304+P340,P304+P341,P305+P351+P338,P312,P330,P332+P313,P337+P313,P342+P311,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-2488746
pictograms GHS07,GHS08
hstatements H302,H312,H315,H319,H332,H334,H335
Availability details for
Quantity
  • Availability
  • Add to Cart