ENAH5802EAFF Enamine

1,1-difluoro-2-(2-isocyanatoethoxy)ethane

  • MDL number MFCD32066724

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7091567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7091567
Availability details for
Quantity
  • Availability
  • Add to Cart