ENAH5802EB01 Enamine

2-isocyanato-1,5-dimethyl-1H-indole

  • MDL number MFCD32198043
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-isocyanato-1,5-dimethyl-1H-indole
Inchi Code 1S/C11H10N2O/c1-8-3-4-10-9(5-8)6-11(12-7-14)13(10)2/h3-6H,1-2H3
Inchi Key SYRSKDUZPULCGD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7443259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443259
Availability details for
Quantity
  • Availability
  • Add to Cart