ENAH5802EB02 Enamine

[(3-isocyanatopropoxy)methyl]benzene

  • MDL number MFCD30212332
  • Cas Number : 163233-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7175938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7175938
Availability details for
Quantity
  • Availability
  • Add to Cart