ENAH5802EB07 Enamine

3-(2-isothiocyanatoethoxy)prop-1-ene

  • Cas Number : 1874289-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7338306

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7338306
Availability details for
Quantity
  • Availability
  • Add to Cart