ENAH5802EB0E Enamine

1-[(1S,2R)-2-methylcyclopropyl]ethan-1-one

  • MDL number MFCD24689672
  • Cas Number : 35625-37-7
  • Molecular Weight : 98.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-((1S,2R)-2-methylcyclopropyl)ethan-1-one
Inchi Code 1S/C6H10O/c1-4-3-6(4)5(2)7/h4,6H,3H2,1-2H3/t4-,6+/m1/s1
Inchi Key CSEAGYCTZLEZON-XINAWCOVSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7071888

Safety Information

signalword Warning
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P271,P280,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7071888
pictograms GHS02,GHS07
hstatements H226,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart