ENAH5802EB0F Enamine

(3S)-3-methylcyclopentan-1-one

  • MDL number MFCD24687931
  • Cas Number : 6672-24-8
  • Molecular Weight : 98.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-3-methylcyclopentan-1-one
Inchi Code 1S/C6H10O/c1-5-2-3-6(7)4-5/h5H,2-4H2,1H3/t5-/m0/s1
Inchi Key AOKRXIIIYJGNNU-YFKPBYRVSA-N

Description

Purity 95
Flash Point 34
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7101944

Safety Information

signalword Warning
pcodes P210,P233,P240,P241,P242,P243,P280,P303+P361+P353,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-7101944
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart