ENAH5802EB10 Enamine

3-(fluoromethyl)cyclobutan-1-one

  • MDL number MFCD24710636
  • Cas Number : 913339-21-6
  • Molecular Weight : 102.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7213929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7213929
Availability details for
Quantity
  • Availability
  • Add to Cart