ENAH5802EB13 Enamine

2-acetylcyclopropane-1-carbonitrile

  • MDL number MFCD24737182
  • Cas Number : 1824580-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7232413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7232413
Availability details for
Quantity
  • Availability
  • Add to Cart