ENAH5802EB14 Enamine

1-acetylcyclopropane-1-carbonitrile

  • MDL number MFCD18633040
  • Cas Number : 146857-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-2960941

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2960941
Availability details for
Quantity
  • Availability
  • Add to Cart