ENAH5802EB19 Enamine

2,2-dimethylpyrrolidin-3-one hydrochloride

  • MDL number MFCD26593757
  • Cas Number : 1554561-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-325793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-325793
Availability details for
Quantity
  • Availability
  • Add to Cart