ENAH5802EB1B Enamine

(2R)-2-fluorocyclohexan-1-one

  • MDL number MFCD32069252
  • Cas Number : 200118-23-6
  • Molecular Weight : 116.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-2-fluorocyclohexan-1-one
Inchi Code 1S/C6H9FO/c7-5-3-1-2-4-6(5)8/h5H,1-4H2/t5-/m1/s1
Inchi Key VQYOFTVCYSPHPG-RXMQYKEDSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7349390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7349390
Availability details for
Quantity
  • Availability
  • Add to Cart