ENAH5802EB1C Enamine

(2S)-2-fluorocyclohexan-1-one

  • MDL number MFCD29987213
  • Cas Number : 918300-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7252105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7252105
Availability details for
Quantity
  • Availability
  • Add to Cart