ENAH5802EB22 Enamine

5-ethylcyclohex-2-en-1-one

  • MDL number MFCD24709626
  • Cas Number : 40790-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7435570

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7435570
Availability details for
Quantity
  • Availability
  • Add to Cart