ENAH5802EB23 Enamine

6-methylhept-1-en-3-one

  • MDL number MFCD21262992
  • Cas Number : 42809-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-6218525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6218525
Availability details for
Quantity
  • Availability
  • Add to Cart