ENAH5802EB28 Enamine

3,3-difluorocyclohexan-1-one

  • MDL number MFCD28015614
  • Cas Number : 1432447-08-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-262621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-262621
Availability details for
Quantity
  • Availability
  • Add to Cart