ENAH5802EB2C Enamine

5,5-difluoro-4-methylpentan-2-one

  • MDL number MFCD32198342
  • Cas Number : 2375262-42-1
  • Molecular Weight : 136.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7120982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7120982
Availability details for
Quantity
  • Availability
  • Add to Cart