ENAH5802EB2D Enamine

4H,5H,6H-cyclopenta[c]thiophen-5-one

  • MDL number MFCD18451698
  • Cas Number : 38447-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6197990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6197990
Availability details for
Quantity
  • Availability
  • Add to Cart