ENAH5802EB34 Enamine

1-(2-oxopropyl)cyclopropane-1-carboxylic acid

  • Cas Number : 1784614-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6308915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6308915
Availability details for
Quantity
  • Availability
  • Add to Cart