ENAH5802EB36 Enamine

1,1-dimethylsilinan-4-one

  • MDL number MFCD24688094
  • Cas Number : 18276-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-6194023

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6194023
Availability details for
Quantity
  • Availability
  • Add to Cart