ENAH5802EB37 Enamine

(3R,4S)-3,4-dimethoxycyclopentan-1-one

  • Cas Number : 2375250-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431378
Availability details for
Quantity
  • Availability
  • Add to Cart