ENAH5802EB3A Enamine

4-(dimethylphosphoryl)butan-2-one

  • MDL number MFCD24701712
  • Cas Number : 53314-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-6754542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6754542
Availability details for
Quantity
  • Availability
  • Add to Cart