ENAH5802EB3B Enamine

(2R,7S)-2,7-difluorocycloheptan-1-one

  • MDL number MFCD32198041
  • Molecular Weight : 148.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2R,7S)-2,7-difluorocycloheptan-1-one

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart