ENAH5802EB40 Enamine

2-tert-butyl-4-[(3,4,5-trimethoxyphenyl)methylidene]-1,2,3,4-tetrahydroacridine-9-carboxylic acid

  • Cas Number : 519150-66-4
  • Molecular Weight : 461.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-221
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-00913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-00913
Availability details for
Quantity
  • Availability
  • Add to Cart