ENAH5802EB41 Enamine

1-(furan-2-yl)-2,2-dimethylpropan-1-one

  • MDL number MFCD16154992
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-furyl)-2,2-dimethyl-1-propanone
Inchi Code 1S/C9H12O2/c1-9(2,3)8(10)7-5-4-6-11-7/h4-6H,1-3H3
Inchi Key KUMBYTKNBWARAD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-108627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108627
Availability details for
Quantity
  • Availability
  • Add to Cart