ENAH5802EB45 Enamine

1-cycloheptylpropan-1-one

  • MDL number MFCD20801147
  • Cas Number : 89932-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1246607

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1246607
Availability details for
Quantity
  • Availability
  • Add to Cart