ENAH5802EB48 Enamine

ethyl (2E)-2-methyl-4-oxopent-2-enoate

  • Cas Number : 126314-15-6
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7426429

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7426429
Availability details for
Quantity
  • Availability
  • Add to Cart