ENAH5802EB4B Enamine

3-methoxy-2,2,4,4-tetramethylcyclobutan-1-one

  • MDL number MFCD24702968
  • Cas Number : 19203-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7432138

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432138
Availability details for
Quantity
  • Availability
  • Add to Cart