ENAH5802EB4D Enamine

2-nitrocycloheptan-1-one

  • MDL number MFCD09865095
  • Cas Number : 13154-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6772348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6772348
Availability details for
Quantity
  • Availability
  • Add to Cart