ENAH5802EB4E Enamine

4-methyl-2-nitrocyclohexan-1-one

  • Cas Number : 81842-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427820
Availability details for
Quantity
  • Availability
  • Add to Cart