ENAH5802EB4F Enamine

2-methoxy-1-(piperidin-3-yl)ethan-1-one hydrochloride

  • MDL number MFCD32066216
  • Cas Number : 2361643-90-3
  • Molecular Weight : 193.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7433796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433796
Availability details for
Quantity
  • Availability
  • Add to Cart