ENAH5802EB51 Enamine

1',3'-dihydrospiro[cyclopropane-1,2'-indene]-3'-one

  • MDL number MFCD21877680
  • Cas Number : 22228-23-5
  • Molecular Weight : 158.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-3035547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3035547
Availability details for
Quantity
  • Availability
  • Add to Cart