ENAH5802EB54 Enamine

3-methyl-1,2,3,4-tetrahydroquinolin-4-one

  • MDL number MFCD20661867
  • Cas Number : 30448-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-7425290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7425290
Availability details for
Quantity
  • Availability
  • Add to Cart